Assay Details
| Gene Symbol: Ap3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009680.3 | NP_033810.2 | 5 - 6 | 674 | 101 | |
| GenBank mRNA | AF103809.2 | - | 5 - 6 | 611 | 101 | |
| AK143559.1 | - | 5 - 6 | 670 | 101 | ||
| AK163957.1 | - | 5 - 6 | 612 | 101 | ||
| AK171595.1 | - | 5 - 6 | 675 | 101 | ||
| BC015068.1 | - | 5 - 6 | 671 | 4018563 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ap3b1
Genomic Map