Assay Details
| Gene Symbol: Arsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009712.3 | NP_033842.3 | 4 - 5 | 997 | 69 | |
| GenBank mRNA | AK083309.1 | - | 4 - 5 | 981 | 69 | |
| AK140163.1 | - | 4 - 5 | 1047 | 69 | ||
| AK154098.1 | - | 4 - 5 | 917 | 69 | ||
| AK158312.1 | - | 4 - 5 | 998 | 69 | ||
| AK190270.1 | - | 1 - 2 | 59 | 69 | ||
| AK205135.1 | - | 1 - 2 | 59 | 69 | ||
| BC150661.1 | - | 4 - 5 | 954 | 9087572 | 69 | |
| X92096.1 | - | 3 - 4 | 490 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arsb
Genomic Map