Assay Details
| Gene Symbol: Scamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029153.1 | NP_083429.1 | 2 - 3 | 139 | 82 | |
| GenBank mRNA | AK135058.1 | - | 2 - 3 | 153 | 82 | |
| AK154671.1 | - | 2 - 3 | 153 | 82 | ||
| AK162168.1 | - | 2 - 3 | 344 | 82 | ||
| AK162198.1 | - | 2 - 3 | 574 | 82 | ||
| AK170343.1 | - | 2 - 3 | 190 | 82 | ||
| BC034283.1 | - | 2 - 3 | 139 | 4162134 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK162168.1 transcripts.
Set Membership: |
|

Scamp1
Genomic Map