Assay Details
| Gene Symbol: Scamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029153.1 | NP_083429.1 | 3 - 4 | 241 | 64 | |
| GenBank mRNA | AK015706.1 | - | 2 - 3 | 202 | 64 | |
| AK132867.1 | - | 2 - 3 | 211 | 64 | ||
| AK135058.1 | - | 3 - 4 | 255 | 64 | ||
| AK154671.1 | - | 3 - 4 | 255 | 64 | ||
| AK162168.1 | - | 3 - 4 | 446 | 64 | ||
| AK162198.1 | - | 3 - 4 | 676 | 64 | ||
| AK170343.1 | - | 3 - 4 | 292 | 64 | ||
| BC034283.1 | - | 3 - 4 | 241 | 4162134 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scamp1
Genomic Map