Assay Details
| Gene Symbol: Scamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029153.1 | NP_083429.1 | 6 - 7 | 638 | 76 | |
| GenBank mRNA | AK015706.1 | - | 5 - 6 | 599 | 76 | |
| AK132867.1 | - | 5 - 6 | 608 | 76 | ||
| AK135058.1 | - | 6 - 7 | 652 | 76 | ||
| AK162168.1 | - | 6 - 7 | 843 | 76 | ||
| AK170343.1 | - | 6 - 7 | 689 | 76 | ||
| BC034283.1 | - | 6 - 7 | 638 | 4162134 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scamp1
Genomic Map