Assay Details
| Gene Symbol: Dmgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028772.3 | NP_083048.1 | 3 - 4 | 365 | 70 | |
| GenBank mRNA | AK004755.1 | - | 3 - 4 | 366 | 70 | |
| BC089599.1 | - | 3 - 4 | 368 | 30118948 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dmgdh
Genomic Map