Assay Details
| Gene Symbol: Magea4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020280.2 | NP_064676.2 | 1 | 51 | 95 | |
| GenBank mRNA | BC104088.1 | - | 1 | 51 | 40042624 | 95 |
| BC104089.1 | - | 1 | 51 | 40042626 | 95 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Magea4
Genomic Map