Assay Details
| Gene Symbol: Gm12227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC067249.1 | - | 1 - 2 | 181 | 65 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rap guanine nucleotide exchange factor (GEF) 6 | Rapgef6 |
A530068K01, AI844632, BB085664, C030018K18Rik, PDZ-GEF2, Pdzgef2, RA-GEF-2, Ragef2, mKIAA4052 |
-29.4kb | Sense | Chr. 11 - 54521754 - 54699286 on NCBI Build GRCm38 |
| Downstream |
LYR motif containing 7, opposite strand |
Lyrm7os |
Gm12228, OTTMUSG00000005674 |
90.8kb | Sense | Chr. 11 - 54819919 - 54824967 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm12227
Genomic Map