Assay Details
| Gene Symbol: Col4a3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023420.2 | NP_075909.1 | 10 - 11 | 1533 | 78 | |
| GenBank mRNA | AF232932.1 | - | 10 - 11 | 1547 | 78 | |
| AK080958.1 | - | 10 - 11 | 1531 | 78 | ||
| BC016197.1 | - | 10 - 11 | 1496 | 4502118 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Col4a3bp
Genomic Map