Assay Details
| Gene Symbol: Col4a3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164222.1 | NP_001157694.1 | 14 - 15 | 1966 | 112 | |
| NM_023420.2 | NP_075909.1 | 15 - 16 | 2044 | 112 | ||
| GenBank mRNA | AF232932.1 | - | 15 - 16 | 2058 | 112 | |
| AF232934.1 | - | 14 - 15 | 1980 | 112 | ||
| AK012989.1 | - | 14 - 15 | 1947 | 112 | ||
| AK018103.1 | - | 3 - 4 | 335 | 112 | ||
| AK030305.1 | - | 15 - 16 | 1874 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Col4a3bp
Genomic Map