Assay Details
| Gene Symbol: Hmgcr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008255.2 | NP_032281.2 | 10 - 11 | 1340 | 72 | |
| GenBank mRNA | AK159899.1 | - | 9 - 10 | 1228 | 72 | |
| BC019782.1 | - | 9 - 10 | 1216 | 72 | ||
| BC085083.1 | - | 10 - 11 | 1252 | 5320486 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hmgcr
Genomic Map