Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300016500.mm.1 |
NM_029210.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016500.mm.1 |
NM_029210.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016500.mm.1 |
TC1300002468.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300016502.mm.1 |
NM_029210.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016502.mm.1 |
NM_029210.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016502.mm.1 |
TC1300002468.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300016499.mm.1 |
NM_029210.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016499.mm.1 |
NM_029210.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016499.mm.1 |
TC1300002468.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300016503.mm.1 |
NM_029210.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016503.mm.1 |
NM_029210.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300016503.mm.1 |
TC1300002468.mm.1 |
MTA TC |