Assay Details
| Gene Symbol: Sv2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029210.1 | NP_083486.1 | 10 - 11 | 1863 | 118 | |
| GenBank mRNA | AK015921.1 | - | 3 - 4 | 368 | 118 | |
| AK137053.1 | - | 6 - 7 | 705 | 118 | ||
| AK173092.1 | - | 10 - 11 | 1863 | 118 | ||
| BC132111.1 | - | 9 - 10 | 1699 | 40130388 | 118 | |
| BC137861.1 | - | 9 - 10 | 1699 | 8860880 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sv2c
Genomic Map