Assay Details
| Gene Symbol: Jmy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021310.3 | NP_067285.2 | 4 - 5 | 2003 | 111 | |
| GenBank mRNA | AB221690.1 | - | 4 - 5 | 1501 | 111 | |
| AF201390.1 | - | 4 - 5 | 1627 | 111 | ||
| AK139934.1 | - | 4 - 5 | 1806 | 111 | ||
| AK141957.1 | - | 4 - 5 | 2004 | 111 | ||
| BC069906.1 | - | 4 - 5 | 1857 | 111 | ||
| BC090835.1 | - | 4 - 5 | 1884 | 5705427 | 111 | |
More Information
Updated Mapping Information
Set Membership: |
|

Jmy
Genomic Map