Assay Details
| Gene Symbol: Papd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133905.2 | NP_598666.1 | 8 - 9 | 1049 | 115 | |
| GenBank mRNA | AK033141.1 | - | 5 - 6 | 384 | 115 | |
| AK146086.1 | - | 8 - 9 | 967 | 115 | ||
| AM419011.1 | - | 7 - 8 | 810 | 115 | ||
| BC016629.1 | - | 8 - 9 | 1049 | 3595512 | 115 | |
More Information
Set Membership: |
|

Papd4
Genomic Map