| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300007091.mm.1 |
NM_001271388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007091.mm.1 |
NM_001271389.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007091.mm.1 |
NM_023472.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007091.mm.1 |
TC1300001112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300007082.mm.1 |
NM_001271388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007082.mm.1 |
NM_001271389.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007082.mm.1 |
NM_001271390.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007082.mm.1 |
NM_001271391.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007082.mm.1 |
NM_001271392.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007082.mm.1 |
NM_023472.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007082.mm.1 |
TC1300001112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300007087.mm.1 |
NM_001271388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007087.mm.1 |
NM_001271389.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007087.mm.1 |
NM_001271390.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007087.mm.1 |
NM_001271391.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007087.mm.1 |
NM_001271392.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007087.mm.1 |
NM_023472.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007087.mm.1 |
TC1300001112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300007083.mm.1 |
NM_001271388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007083.mm.1 |
NM_001271389.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007083.mm.1 |
NM_001271390.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007083.mm.1 |
NM_001271391.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007083.mm.1 |
NM_001271392.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007083.mm.1 |
NM_023472.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007083.mm.1 |
TC1300001112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300007085.mm.1 |
NM_001271388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007085.mm.1 |
NM_001271389.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007085.mm.1 |
NM_001271390.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007085.mm.1 |
NM_001271391.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007085.mm.1 |
NM_001271392.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007085.mm.1 |
NM_023472.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007085.mm.1 |
TC1300001112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300007089.mm.1 |
NM_001271388.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007089.mm.1 |
NM_001271389.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007089.mm.1 |
NM_023472.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300007089.mm.1 |
TC1300001112.mm.1 |
MTA TC |