Assay Details
| Gene Symbol: Ankra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271388.1 | NP_001258317.1 | 5 - 6 | 1109 | 75 | |
| NM_001271389.1 | NP_001258318.1 | 5 - 6 | 1109 | 75 | ||
| NM_001271390.1 | NP_001258319.1 | 4 - 5 | 727 | 75 | ||
| NM_001271391.1 | NP_001258320.1 | 4 - 5 | 727 | 75 | ||
| NM_001271392.1 | NP_001258321.1 | 4 - 5 | 727 | 75 | ||
| NM_023472.2 | NP_075961.1 | 5 - 6 | 1109 | 75 | ||
| GenBank mRNA | AF314031.1 | - | 5 - 6 | 943 | 75 | |
| AK003438.2 | - | 4 - 5 | 646 | 75 | ||
| AK054037.1 | - | 5 - 6 | 915 | 75 | ||
| AK081695.1 | - | 4 - 5 | 612 | 75 | ||
| AK139343.1 | - | 5 - 6 | 991 | 75 | ||
| BC037437.1 | - | 4 - 5 | 655 | 75 | ||
| BC066113.1 | - | 4 - 5 | 646 | 30361246 | 75 | |
| BC079615.1 | - | 5 - 6 | 926 | 30361702 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ankra2
Genomic Map