Assay Details
| Gene Symbol: Pik3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024955.2 | NP_001020126.1 | 9 - 10 | 1515 | 78 | |
| NM_001077495.2 | NP_001070963.1 | 15 - 16 | 2604 | 78 | ||
| GenBank mRNA | AK046259.1 | - | 9 - 10 | 1518 | 78 | |
| AK164783.1 | - | 1 - 2 | 131 | 78 | ||
| BC026146.1 | - | 15 - 16 | 2585 | 3979333 | 78 | |
| BC051106.1 | - | 9 - 10 | 1215 | 5031261 | 78 | |
| M60651.1 | - | 15 - 16 | 2539 | 78 | ||
| U50413.1 | - | 15 - 16 | 2557 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r1
Genomic Map