Assay Details
| Gene Symbol: Pik3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077495.2 | NP_001070963.1 | 1 - 2 | 243 | 86 | |
| GenBank mRNA | AK160493.1 | - | 1 - 2 | 217 | 86 | |
| AK171366.1 | - | 1 - 2 | 241 | 86 | ||
| BC026146.1 | - | 1 - 2 | 224 | 3979333 | 86 | |
| CF169061.1 | - | 1 - 2 | 243 | 86 | ||
| M60651.1 | - | 1 - 2 | 178 | 86 | ||
| U50413.1 | - | 1 - 2 | 196 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC026146.1, M60651.1, NM_001077495.2, U50413.1 transcripts.
Set Membership: |
|

Pik3r1
Genomic Map