Assay Details
| Gene Symbol: Pik3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077495.2 | NP_001070963.1 | 4 - 5 | 1127 | 70 | |
| GenBank mRNA | BB647657.1 | - | 3 - 4 | 344 | 70 | |
| BC026146.1 | - | 4 - 5 | 1108 | 3979333 | 70 | |
| M60651.1 | - | 4 - 5 | 1062 | 70 | ||
| U50413.1 | - | 4 - 5 | 1080 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r1
Genomic Map