Assay Details
| Gene Symbol: Pik3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077495.2 | NP_001070963.1 | 6 - 7 | 1454 | 101 | |
| GenBank mRNA | BC026146.1 | - | 6 - 7 | 1435 | 3979333 | 101 |
| M60651.1 | - | 6 - 7 | 1389 | 101 | ||
| U50413.1 | - | 6 - 7 | 1407 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r1
Genomic Map