Assay Details
| Gene Symbol: Slc30a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022885.2 | NP_075023.2 | 1 - 2 | 324 | 137 | |
| GenBank mRNA | AF461761.1 | - | 1 - 2 | 223 | 137 | |
| AK051183.1 | - | 1 - 2 | 324 | 137 | ||
| AK088004.1 | - | 1 - 2 | 239 | 137 | ||
| AK090176.1 | - | 1 - 2 | 233 | 137 | ||
| AK166006.1 | - | 1 - 2 | 134 | 137 | ||
| BC033452.1 | - | 1 - 2 | 223 | 3156622 | 137 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF461761.1, AK051183.1, AK088004.1, AK166006.1, BC033452.1, NM_022885.2 transcripts.
Set Membership: |
|

Slc30a5
Genomic Map