Assay Details
| Gene Symbol: Slc30a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022885.2 | NP_075023.2 | 3 - 4 | 513 | 119 | |
| GenBank mRNA | AF461761.1 | - | 3 - 4 | 412 | 119 | |
| AK051183.1 | - | 3 - 4 | 513 | 119 | ||
| AK088004.1 | - | 3 - 4 | 428 | 119 | ||
| AK090176.1 | - | 3 - 4 | 422 | 119 | ||
| AK166006.1 | - | 3 - 4 | 323 | 119 | ||
| BC033452.1 | - | 3 - 4 | 412 | 3156622 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a5
Genomic Map