Assay Details
| Gene Symbol: Slc30a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022885.2 | NP_075023.2 | 9 - 10 | 1320 | 77 | |
| GenBank mRNA | AF233321.1 | - | 2 - 3 | 77 | ||
| AF461761.1 | - | 9 - 10 | 1219 | 77 | ||
| AK051183.1 | - | 9 - 10 | 1320 | 77 | ||
| AK088004.1 | - | 8 - 9 | 1064 | 77 | ||
| AK090176.1 | - | 9 - 10 | 1229 | 77 | ||
| AK166006.1 | - | 9 - 10 | 1130 | 77 | ||
| AK180687.1 | - | 1 - 2 | 51 | 77 | ||
| BC029033.1 | - | 6 - 7 | 762 | 77 | ||
| BC033452.1 | - | 9 - 10 | 1219 | 3156622 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a5
Genomic Map