Assay Details
| Gene Symbol: Mrps36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190264.1 | NP_001177193.1 | 1 - 2 | 130 | 96 | |
| NM_025369.3 | NP_079645.1 | 1 - 2 | 130 | 96 | ||
| GenBank mRNA | AK003111.1 | - | 1 - 2 | 60 | 96 | |
| AK003765.1 | - | 1 - 2 | 107 | 96 | ||
| AK007512.1 | - | 1 - 2 | 101 | 96 | ||
| AK009050.1 | - | 1 - 2 | 109 | 96 | ||
| AK009966.1 | - | 1 - 2 | 61 | 96 | ||
| BC028499.1 | - | 1 - 2 | 86 | 1382215 | 96 | |
| BC096576.1 | - | 1 - 2 | 95 | 1401014 | 96 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK003111.1, AK003765.1, AK007512.1, AK009050.1, AK009966.1, BC028499.1, BC096576.1, NM_001190264.1, NM_025369.3 transcripts.
Set Membership: |
|

Mrps36
Genomic Map