Assay Details
| Gene Symbol: Fcho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172591.3 | NP_766179.2 | 20 - 21 | 2021 | 82 | |
| GenBank mRNA | AK031041.1 | - | 20 - 21 | 2024 | 82 | |
| AK142282.1 | - | 20 - 21 | 1707 | 82 | ||
| AK161093.1 | - | 1 - 2 | 754 | 82 | ||
| AK164135.1 | - | 21 - 22 | 1879 | 82 | ||
| AK169439.1 | - | 4 - 5 | 1443 | 82 | ||
| BC052456.1 | - | 20 - 21 | 1718 | 6830607 | 82 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK161093.1 transcripts.
Set Membership: |
|

Fcho2
Genomic Map