Assay Details
| Gene Symbol: Fcho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172591.3 | NP_766179.2 | 21 - 22 | 2184 | 141 | |
| GenBank mRNA | AK031041.1 | - | 21 - 22 | 2187 | 141 | |
| AK142282.1 | - | 21 - 22 | 1870 | 141 | ||
| AK161093.1 | - | 2 - 3 | 917 | 141 | ||
| AK164135.1 | - | 22 - 23 | 2042 | 141 | ||
| AK169439.1 | - | 5 - 6 | 1606 | 141 | ||
| BC052456.1 | - | 21 - 22 | 1881 | 6830607 | 141 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK161093.1 transcripts.
Set Membership: |
|

Fcho2
Genomic Map