| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300016875.mm.1 |
NM_172591.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016875.mm.1 |
NM_172591.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016875.mm.1 |
TC1300002507.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016876.mm.1 |
NM_172591.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016876.mm.1 |
NM_172591.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016876.mm.1 |
TC1300002507.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300016874.mm.1 |
NM_172591.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016874.mm.1 |
NM_172591.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300016874.mm.1 |
TC1300002507.mm.1 |
MTA TC |