Assay Details
| Gene Symbol: Fcho2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172591.3 | NP_766179.2 | 3 - 4 | 537 | 161 | |
| GenBank mRNA | AK031041.1 | - | 3 - 4 | 540 | 161 | |
| AK142282.1 | - | 3 - 4 | 223 | 161 | ||
| AK146566.1 | - | 3 - 4 | 271 | 161 | ||
| AK164135.1 | - | 3 - 4 | 283 | 161 | ||
| BC019933.1 | - | 4 - 5 | 273 | 161 | ||
| BC047122.1 | - | 4 - 5 | 273 | 161 | ||
| BC052456.1 | - | 3 - 4 | 234 | 6830607 | 161 | |
| BC053718.1 | - | 3 - 4 | 253 | 161 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK164135.1 transcripts.
Set Membership: |
|

Fcho2
Genomic Map