Assay Details
| Gene Symbol: Havcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166631.1 | NP_001160103.1 | 5 - 6 | 769 | 98 | |
| NM_001166632.1 | NP_001160104.1 | 5 - 6 | 653 | 98 | ||
| NM_134248.2 | NP_599009.2 | 5 - 6 | 677 | 98 | ||
| GenBank mRNA | AF399829.1 | - | 5 - 6 | 668 | 98 | |
| AF399830.1 | - | 4 - 5 | 599 | 98 | ||
| AK133466.1 | - | 5 - 6 | 771 | 98 | ||
| BC053400.1 | - | 5 - 6 | 661 | 30033601 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Havcr1
Genomic Map