Assay Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002004.2 | NP_001002004.2 | 10 - 11 | 1178 | 66 | |
| GenBank mRNA | AK149719.1 | - | 10 - 11 | 1149 | 66 | |
| AK150829.1 | - | 10 - 11 | 1179 | 66 | ||
| BC019798.1 | - | 10 - 11 | 1131 | 66 | ||
| BC075633.1 | - | 10 - 11 | 1138 | 30537398 | 66 | |
| BC080706.1 | - | 10 - 11 | 1155 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

2610507B11Rik
Genomic Map