Assay Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002004.2 | NP_001002004.2 | 13 - 14 | 1609 | 76 | |
| GenBank mRNA | AK149719.1 | - | 13 - 14 | 1580 | 76 | |
| BC075633.1 | - | 13 - 14 | 1569 | 30537398 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

2610507B11Rik
Genomic Map