Assay Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002004.2 | NP_001002004.2 | 14 - 15 | 1867 | 78 | |
| GenBank mRNA | AK012059.1 | - | 1 - 2 | 138 | 78 | |
| AK149719.1 | - | 14 - 15 | 1838 | 78 | ||
| BC075633.1 | - | 14 - 15 | 1827 | 30537398 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK012059.1 transcripts.
Set Membership: |
|

2610507B11Rik
Genomic Map