Assay Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002004.2 | NP_001002004.2 | 15 - 16 | 2072 | 107 | |
| GenBank mRNA | AK012059.1 | - | 2 - 3 | 343 | 107 | |
| BC075633.1 | - | 15 - 16 | 2032 | 30537398 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

2610507B11Rik
Genomic Map