Assay Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002004.2 | NP_001002004.2 | 16 - 17 | 3166 | 63 | |
| GenBank mRNA | AK122206.1 | - | 1 - 2 | 818 | 63 | |
| BC040464.1 | - | 1 - 2 | 560 | 63 | ||
| BC075633.1 | - | 16 - 17 | 3126 | 30537398 | 63 | |
| X81632.1 | - | 1 - 2 | 1075 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

2610507B11Rik
Genomic Map