Assay Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002004.2 | NP_001002004.2 | 20 - 21 | 3900 | 105 | |
| GenBank mRNA | AK122206.1 | - | 5 - 6 | 1552 | 105 | |
| BC040464.1 | - | 5 - 6 | 1294 | 105 | ||
| BC075633.1 | - | 20 - 21 | 3860 | 30537398 | 105 | |
| X81632.1 | - | 5 - 6 | 1809 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

2610507B11Rik
Genomic Map