Assay Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002004.2 | NP_001002004.2 | 22 - 23 | 4196 | 73 | |
| GenBank mRNA | AK122206.1 | - | 7 - 8 | 1848 | 73 | |
| BC040464.1 | - | 7 - 8 | 1590 | 73 | ||
| BC075633.1 | - | 22 - 23 | 4156 | 30537398 | 73 | |
| X81632.1 | - | 7 - 8 | 2105 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

2610507B11Rik
Genomic Map