Assay Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002004.2 | NP_001002004.2 | 2 - 3 | 295 | 107 | |
| GenBank mRNA | AK149719.1 | - | 2 - 3 | 266 | 107 | |
| AK150829.1 | - | 2 - 3 | 296 | 107 | ||
| BB866513.1 | - | 2 - 3 | 295 | 107 | ||
| BC019798.1 | - | 2 - 3 | 107 | |||
| BC080706.1 | - | 2 - 3 | 272 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

2610507B11Rik
Genomic Map