Assay Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002004.2 | NP_001002004.2 | 4 - 5 | 492 | 82 | |
| GenBank mRNA | AK149719.1 | - | 4 - 5 | 463 | 82 | |
| AK150829.1 | - | 4 - 5 | 493 | 82 | ||
| BB866513.1 | - | 4 - 5 | 492 | 82 | ||
| BC019798.1 | - | 4 - 5 | 445 | 82 | ||
| BC075633.1 | - | 4 - 5 | 452 | 30537398 | 82 | |
| BC080706.1 | - | 4 - 5 | 469 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

2610507B11Rik
Genomic Map