Assay Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002004.2 | NP_001002004.2 | 5 - 6 | 611 | 104 | |
| GenBank mRNA | AK149719.1 | - | 5 - 6 | 582 | 104 | |
| AK150829.1 | - | 5 - 6 | 612 | 104 | ||
| BC019798.1 | - | 5 - 6 | 564 | 104 | ||
| BC075633.1 | - | 5 - 6 | 571 | 30537398 | 104 | |
| BC080706.1 | - | 5 - 6 | 588 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

2610507B11Rik
Genomic Map