Assay Details
| Gene Symbol: 2610507B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002004.2 | NP_001002004.2 | 7 - 8 | 854 | 64 | |
| GenBank mRNA | AK149719.1 | - | 7 - 8 | 825 | 64 | |
| AK150829.1 | - | 7 - 8 | 855 | 64 | ||
| BC019798.1 | - | 7 - 8 | 807 | 64 | ||
| BC075633.1 | - | 7 - 8 | 814 | 30537398 | 64 | |
| BC080706.1 | - | 7 - 8 | 831 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

2610507B11Rik
Genomic Map