Assay Details
| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009012.2 | NP_033038.2 | 2 - 3 | 476 | 118 | |
| GenBank mRNA | AK018001.1 | - | 2 - 3 | 476 | 118 | |
| AK037641.1 | - | 2 - 3 | 477 | 118 | ||
| AK144155.1 | - | 2 - 3 | 468 | 118 | ||
| BC058180.1 | - | 2 - 3 | 452 | 118 | ||
| U66887.1 | - | 2 - 3 | 454 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rad50
Genomic Map