Assay Details
| Gene Symbol: Rad50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009012.2 | NP_033038.2 | 6 - 7 | 1141 | 119 | |
| GenBank mRNA | AK037641.1 | - | 6 - 7 | 1142 | 119 | |
| AK144155.1 | - | 6 - 7 | 1133 | 119 | ||
| BC058180.1 | - | 6 - 7 | 1117 | 119 | ||
| U66887.1 | - | 6 - 7 | 1119 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rad50
Genomic Map