Assay Details
| Gene Symbol: Ifi203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF022371.1 | - | 738 | 61 | ||
| AK040214.1 | - | 5 - 6 | 678 | 61 | ||
| AK161531.1 | - | 3 - 4 | 521 | 61 | ||
| AK164695.1 | - | 3 - 4 | 515 | 61 | ||
| AK169754.1 | - | 5 - 6 | 672 | 61 | ||
| BE632791.1 | - | 3 - 4 | 456 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK164695.1 transcripts.
Set Membership: |
|

Ifi203
Genomic Map