Assay Details
| Gene Symbol: Pde8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170669.1 | NP_001164140.1 | 12 - 13 | 1525 | 63 | |
| NM_172263.2 | NP_758467.2 | 13 - 14 | 1258 | 63 | ||
| GenBank mRNA | AK036090.1 | - | 15 - 16 | 1595 | 63 | |
| AK043680.1 | - | 12 - 13 | 1518 | 63 | ||
| AK045991.1 | - | 12 - 13 | 1527 | 63 | ||
| AK083166.1 | - | 12 - 13 | 1163 | 63 | ||
| AK138301.1 | - | 2 - 3 | 192 | 63 | ||
| BC032286.1 | - | 13 - 14 | 1270 | 5367055 | 63 | |
| EF644173.1 | - | 15 - 16 | 1270 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK138301.1 transcripts.
Set Membership: |
|

Pde8b
Genomic Map