Assay Details
| Gene Symbol: Crhbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198408.3 | NP_940800.1 | 2 - 3 | 326 | 91 | |
| GenBank mRNA | AK135067.1 | - | 2 - 3 | 327 | 91 | |
| AK139404.1 | - | 2 - 3 | 172 | 91 | ||
| BC061247.1 | - | 2 - 3 | 267 | 30246702 | 91 | |
More Information
Set Membership: |
|

Crhbp
Genomic Map