Assay Details
| Gene Symbol: Crhbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198408.3 | NP_940800.1 | 4 - 5 | 700 | 82 | |
| GenBank mRNA | AK135067.1 | - | 4 - 5 | 701 | 82 | |
| AK139404.1 | - | 4 - 5 | 546 | 82 | ||
| BC061247.1 | - | 4 - 5 | 641 | 30246702 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Crhbp
Genomic Map