Assay Details
| Gene Symbol: Crhbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198408.3 | NP_940800.1 | 6 - 7 | 963 | 62 | |
| GenBank mRNA | AK135067.1 | - | 6 - 7 | 964 | 62 | |
| AK139404.1 | - | 6 - 7 | 809 | 62 | ||
| BC061247.1 | - | 6 - 7 | 904 | 30246702 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
synaptic transmission, dopaminergic
maternal aggressive behavior
inflammatory response
female pregnancy
hormone-mediated signaling pathway
cellular response to stress
cellular response to drug
cellular response to potassium ion
hormone metabolic process
regulated exocytosis
behavioral response to ethanol
regulation of corticotropin secretion
negative regulation of corticotropin secretion
cellular response to calcium ion
cellular response to cocaine
cellular response to cAMP
cellular response to tumor necrosis factor
cellular response to estrogen stimulus
cellular response to estradiol stimulus
cellular response to gonadotropin-releasing hormone
negative regulation of corticotropin-releasing hormone receptor activity
regulation of N-methyl-D-aspartate selective glutamate receptor activity
peptide binding
corticotropin-releasing hormone binding
synaptic transmission, dopaminergic
maternal aggressive behavior
inflammatory response
female pregnancy
hormone-mediated signaling pathway
cellular response to stress
cellular response to drug
cellular response to potassium ion
hormone metabolic process
regulated exocytosis
behavioral response to ethanol
regulation of corticotropin secretion
negative regulation of corticotropin secretion
cellular response to calcium ion
cellular response to cocaine
cellular response to cAMP
cellular response to tumor necrosis factor
cellular response to estrogen stimulus
cellular response to estradiol stimulus
cellular response to gonadotropin-releasing hormone
negative regulation of corticotropin-releasing hormone receptor activity
regulation of N-methyl-D-aspartate selective glutamate receptor activity
peptide binding
corticotropin-releasing hormone binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P460734 | BC061247.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P460734 | BC061247.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410192 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713481 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 915100 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 907836 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561210 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 770120 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 959610 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 178228 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155927 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157749 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P460734 | BC061247.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P460734 | BC061247.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240743 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240735 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240758 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240745 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240740 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240751 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240755 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240750 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240730 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240752 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240753 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240756 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240759 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240749 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240747 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P29240722 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155927 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157749 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07960213 | - | - |
| Whole Mouse Genome G4122A | A_52_P460734 | BC061247.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P460734 | BC061247.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112178 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157749 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155927 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10523297 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10523294 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10523292 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10523284 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10523299 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10523286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240769 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240782 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240728 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240776 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29240687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10613295 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240694 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240735 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240709 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240702 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240704 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240760 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240745 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240610 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240697 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10613294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240777 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240604 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240599 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240693 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240728 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240747 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240776 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240758 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10640725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240698 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240753 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240772 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240737 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11051764 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240718 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240608 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240769 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240602 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240711 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10523286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240714 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240767 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10789585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10636061 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240773 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10789588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10523281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10523297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240722 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29240764 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10411218 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411220 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411221 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411222 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411219 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411217 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10411216 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436127_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10411215 | - | - |
| Mouse Genome 430 2.0 Array | 1436127_at | - | - |
| Mouse Genome 430 2.0 Array | 1436127_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12919_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12919_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300016423.mm.1 | NM_198408.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300016423.mm.1 | NM_198408.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300016423.mm.1 | TC1300002458.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436127_PM_at | - | - |
| Mouse Expression Set 430 | 1436127_at | - | - |
| Murine Genome U74v2 Set | 136218_at | - | - |
| Murine Genome U74v2 Set | 136218_PM_at | - | - |
| Murine Genome U74v2 Set | 136270_at | - | - |
| Murine Genome U74v2 Set | 136270_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436127_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436127_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 472648 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5420181 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5420181 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005420181 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231710 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231710 | - | - |
| mouse-6 v1.1 (Illumina) | scl012919.2_18-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5420181 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl012919.2_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5420181 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1231710 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231710 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl012919.2_18-S | - | - |

Crhbp
Genomic Map