Assay Details
| Gene Symbol: Aggf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025630.3 | NP_079906.2 | 3 - 4 | 814 | 86 | |
| GenBank mRNA | AK008168.1 | - | 3 - 4 | 787 | 86 | |
| AK008174.1 | - | 3 - 4 | 784 | 86 | ||
| AK008399.1 | - | 3 - 4 | 787 | 86 | ||
| AK017248.1 | - | 3 - 4 | 770 | 86 | ||
| AK132575.1 | - | 3 - 4 | 815 | 86 | ||
| AY500995.1 | - | 3 - 4 | 787 | 86 | ||
| BC052410.1 | - | 3 - 4 | 791 | 5716440 | 86 | |
More Information
Set Membership: |
|

Aggf1
Genomic Map