Assay Details
| Gene Symbol: Aggf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025630.3 | NP_079906.2 | 9 - 10 | 1755 | 64 | |
| GenBank mRNA | AK009533.1 | - | 5 - 6 | 718 | 64 | |
| AK132575.1 | - | 9 - 10 | 1756 | 64 | ||
| AY500995.1 | - | 9 - 10 | 1728 | 64 | ||
| BC027286.1 | - | 4 - 5 | 431 | 64 | ||
| BC052410.1 | - | 9 - 10 | 1732 | 5716440 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aggf1
Genomic Map